Human umbilical vein endothelial cells (HUVEC), PromoCell
Supplier: PromoCell
C-12255 C-12203 C-14009 C-14012 C-12208 C-14011 C-12250 C-12206 C-12209 C-12207 C-12253 C-12205 C-12200 C-14010 C-14008
10175-200EA 273.17 CAD
10175-200 10175-196 10175-198 10175-192 10171-962 10171-910 10175-194 10171-920 10171-960 10171-916 10171-918 10171-906 10171-956 10171-912 10171-914
Human umbilical vein endothelial cells (HUVEC), PromoCell
Cells
Primary Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from the vein of the umbilical cord and are commonly used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, angiogenesis, and fibrinolysis.
Ordering information: PromoCell offers HUVEC isolated in standard Endothelial Cell Growth Medium and also in Endothelial Cell Growth Medium 2. The cells are supplied either from single donors or from pooled donors (from up to four different umbilical cords). Human Umbilical Artery Endothelial Cells (HUAEC) from the same donor are available on request. In addition, HUVEC are available which are pre-screened for VEGF (vascular endothelial growth factor) response. VEGF is an important signaling protein involved in both vasculogenesis and angiogenesis. In vitro, VEGF has been shown to stimulate endothelial cell mitogenesis, cell migration, sprouting, and microvascular permeability.