Human Cardiac Fibroblasts (HCF), PromoCell
Supplier: Promocell
C-12375 C-14036
10171-994EA 3028.01 CAD
10171-994 10171-996 10172-208
Human Cardiac Fibroblasts (HCF), PromoCell
Cells
Primary Human Cardiac Fibroblasts (HCF) are isolated from the ventricles of an adult heart
  • CD90 positive
  • Cryopreserved at second passage
  • Available from different tissues
This product can be used in the following applications. Wound healing research, Tissue engineering research, Regeneration studies, Toxicological research, Oncology, Epithelial-mesenchymal interaction studies.
They play a central role in the maintenance of the extracellular matrix in the normal heart and the synthesis of growth factors and cytokines. Under pathophysiological conditions, cardiac fibroblasts are involved in scar formation after myocardical infarction, cardiac fibrosis, and cardiac hypertrophy. HCF can be used to study such processes in vitro.
Ordering information: Human Cardiac Fibroblasts can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.