Human Uterine Microvascular Endothelial Cells (HUtMEC), PromoCell
Supplier: Promocell
C-12295 C-14028
10171-986EA 1821.44 CAD
10171-986 10171-988 10172-196
Human Uterine Microvascular Endothelial Cells (HUtMEC), PromoCell
Cells
Primary Human Uterine Microvascular Endothelial Cells (HUtMEC) are isolated from the myometrium (middle layer of the uterine wall) from a single donor who is not pre-treated with hormones. The cells are routinely analyzed by immunofluorescent staining: they stain positive for CD31 and von Willebrand factor and negative for smooth muscle alpha-actin.
During pregnancy HUtMEC propagate intensely and change their phenotype and expression pattern in response to sex steroids
This product can be used in the following applications. Angiogenesis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
This includes up-regulation of HUtMEC nitric oxide synthase activity, modulation of adhesion molecule expression, and an increase in endothelial cell survival and angiogenic activity. HUtMEC are involved in medical conditions like uterine fibroma or adeno-mysis.
Ordering information: Human Human Uterine Microvascular Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.