Human Pulmonary Artery Smooth Muscle Cells (HPASMC), PromoCell
Supplier: Promocell
C-12521 C-14051
10172-032EA 1926.81 CAD
10172-032 10172-034 10172-222
Human Pulmonary Artery Smooth Muscle Cells (HPASMC), PromoCell
Cells
Primary Human Pulmonary Artery Smooth Muscle Cells (HPASMC) are isolated from human pulmonary arteries and stain positive for smooth muscle α-actin
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
They are suited for studying the function of pulmonary arteries under normal and disease conditions (e.g. vasoconstriction induced by alveolar hypoxia). They are also useful for physiological studies of vascular smooth muscle cells.
Ordering information: Human Pulmonary Artery Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.