Human Aortic Adventitial Fibroblasts (HAoAF), PromoCell
Supplier: Promocell
C-12380 C-14037
10171-998EA 1595.06 CAD
10171-998 10172-000 10172-210
Human Aortic Adventitial Fibroblasts (HAoAF), PromoCell
Cells
Primary Human Aortic Adventitial Fibroblasts (HAoAF) are isolated from human aortic adventitial tissue
  • CD90 positive
  • Cryopreserved at second passage
  • Available from different tissues
This product can be used in the following applications. Wound healing research, Tissue engineering research, Regeneration studies, Toxicological research, Oncology, Epithelial-mesenchymal interaction studies
HAoAF can be used for studies concerning vascular diseases e.g. neointimal formation, vascular remodeling, atherosclerosis, and hypertension. They are also often used for the examination of fibroblast disorders like fibrosis or other diseases linked to either imperfect or excessive accumulation of fibroblasts.
Ordering information: Human Aortic Adventitial Fibroblasts can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.