Human Coronary Artery Smooth Muscle Cells (HCASMC), PromoCell
Supplier: Promocell
C-12511 C-14052
10172-028EA 2086.72 CAD
10172-028 10172-030 10172-224
Human Coronary Artery Smooth Muscle Cells (HCASMC), PromoCell
Cells
Primary Human Coronary Artery Smooth Muscle Cells (HCASMC) are isolated from human coronary arteries and stain positive for smooth muscle α-actin. Coronary arteries supply the heart with blood and obstruction in atherosclerosis causes myocardial infarction.
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
HCASMC are qualified to study the function of smooth muscle cells in the development of atherosclerotic plaques or in other diseases like hypertension
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
They are also useful for stent-graft compatibility testing.
Ordering information: Human Coronary Artery Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.