Human Aortic Smooth Muscle Cells (HAoSMC), PromoCell
Supplier: Promocell
Primary Human Aortic Smooth Muscle Cells (HAoSMC) are isolated from plaque-free regions of the human aorta and stain positive for smooth muscle α-actin. The pulsatile pressure produced by the heart causes a cyclic distention of the aorta. The smooth muscle cells in the aortic wall subsequently contract it again. Changes in the arterial wall, associated with vascular diseases such as atherosclerosis and hypertension, strongly influence this process.
- Smooth muscle alpha-actin positive
- CD90 negative
- Routinely morphology tested
- Cryopreserved at second passage
HAoSMC are suitable for studying the role of smooth muscle cells under normal or disease conditions in vitro.
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
Ordering information: Human Aortic Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.