Normal Human Dermal Fibroblasts (NHDF), PromoCell
Supplier: Promocell
C-14030 C-14031 C-12300 C-12302
10172-200EA 302.51 CAD
10172-200 10172-202 10175-136 10175-138 10175-140 10175-142
Normal Human Dermal Fibroblasts (NHDF), PromoCell
Cells
Primary Normal Human Dermal Fibroblasts (NHDF) are isolated form the dermis of juvenile foreskin or adult skin from different locations like the face, the breasts, the abdomen, and the thighs.
  • CD90 positive
  • Cryopreserved at second passage
  • Available from different tissues
Normal Human Epidermal Keratinocytes (NHEK) and Normal Human Epidermal Melanocytes (NHEM) cultured in M2 medium from the same donor are available on request.) They can be used for wound healing studies and dermatological research to investigate diseases like scleroderma, fibrosarcoma, fibrosis, xeroderma pigmentosum, and histiocytoma
This product can be used in the following applications. Wound healing research, Tissue engineering research, Regeneration studies, Toxicological research, Oncology, Epithelial-mesenchymal interaction studies.
Moreover, fibroblasts are important for cancer research, tissue regeneration, and tissue engineering studies.
Ordering information: Normal Human Dermal Fibroblasts can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.