Osteoblast Growth Medium, PromoCell
Supplier: Promocell
C-27001 C-27010 C-27015
10172-338EA 383.51 CAD
10172-338 10172-340 10172-342
Osteoblast Growth Medium, PromoCell
Cell Culture Media
PromoCell Osteoblast Growth Medium was developed to establish and maintain human osteoblast cultures
The fetal calf serum in the medium is strictly pretested for optimal performance. The media are optimized for primary human cells but can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.
Ordering information: The Media (Ready-to-use) consist of one bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Medium. Basal Medium (with or without phenol red) as well as the Osteoblast Growth Medium SupplementMix can also be purchased separately.