Human Dermal Blood Endothelial Cells (HDBEC), PromoCell
Supplier: PromoCell
C-12226 C-14018 C-12211 C-12225 C-12214 C-14019
10172-178EA 382.23 CAD
10172-178 10172-176 10171-928 10171-944 10171-946 10171-924
Human Dermal Blood Endothelial Cells (HDBEC), PromoCell
Cells
Primary Human Dermal Blood Endothelial Cells (HDBEC) are a subpopulation of the Human Dermal Endothelial Cells. They are isolated from the dermis of juvenile foreskin and adult skin (different locations) from a single donor. (Human Dermal Lymphatic Endothelial Cells (HDLEC) from the same donor are available on request.) The cells are routinely analyzed by flow cytometric analysis and immunofluorescent staining: they are > 95% CD31 positive and stain positive for von Willebrand factor. In addition, they are > 90% negative for podoplanin and stain negative for smooth muscle specific alpha-actin.
Blood Endothelial Cells have a key function in physiological processes like vessel tonus, capillary permeability, blood coagulation, fibrolysis, and angiogenesis
This product can be used in the following applications. Angiogenesis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
They are also useful for disease studies of atherosclerosis, tumor genesis, and thrombosis.
Ordering information: Human Dermal Blood Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.