Human Renal Cortical Epithelial Cells (HRCEpC), PromoCell
Supplier: Promocell
Primary Human Renal Cortical Epithelial Cells (HRCEpC) are isolated from the cortex of the human kidney and stain positive for cytokeratin.
- Cytokeratin positive
- Routinely morphology checked
- Cryopreserved at second passage
The renal cortex is the outer portion of the kidney between the renal capsule and the renal medulla
This product can be used in the following applications. Drug uptake studies, Toxicological research, Wound healing research, Oncology, Cell differentiation studies, Respiratory disease research.
It contains the renal corpuscles, the proximal and distal convoluted tubules, and the cortical collecting ducts. Important physiological processes like osmoregulation and excretion can be studied with HRCEpC in vitro.
Ordering information: Human Renal Cortical Epithelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.