New Brunswick™ Premium and HEF® Back-up Systems
Supplier: Eppendorf
U9043-0002 U9043-0006 U9044-0002 U9044-0006
CA97048-924EA 3677.6 CAD
CA97048-924 CA97048-926 CA97048-928 CA97048-930
New Brunswick™ Premium and HEF® Back-up Systems
Freezers Laboratory / General Purpose Freezers
Freezer back-up systems are equipped with a battery back-up to temporarily protect the contents of your freezer in case of power failure
Both systems can be factory installed with your freezer order at no additional charge or retrofitted on location. The CO₂ and LN₂ systems are not interchangeable systems.
Available in either liquid CO₂ and LN₂. Liquid CO₂ can maintain temperatures between −50°C and −70°C, where as LN₂ can go down to −85°C.