Anti-CTLA4 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: UC10-4F10-11]

Supplier: Tonbo Biosciences
60-1522-U025 60-1522-U100
10208-468EA 93.22 CAD
10208-468 10208-470
Anti-CTLA4 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: UC10-4F10-11]
Antibodies
The UC10-4F10-11 antibody is specific for mouse CD152, commonly known as CTLA-4, a 33-37 kDa protein expressed as a homodimer on the surface of activated T and B cells, and on thymocytes. CTLA-4 is structurally similar, yet functionally disparate, to the T cell co-stimulatory molecule CD28. Both CTLA-4 and CD28 interact with the co-stimulatory molecules CD80 (B7-1) and CD86 (B7-2) on antigen-presenting cells, with CTLA-4 displaying a higher avidity than CD28. While CD28 typically delivers a potent co-stimulatory signal in support of T cell activation, CTLA-4 appears to act as a negative regulator of T cell activation and may contribute to the suppressor function of Treg cells.

CTLA-4 proteins may be initially sequestered within Golgi vesicles, from which they can be rapidly transferred to and from the cell surface, a mechanism by which Treg cells can selectively impart suppressive functions. The UC10-4F10-11 antibody may be used for flow cytometric analysis of CTLA-4 expression.

Type: Primary
Antigen: CTLA4
Clonality: Monoclonal
Clone: UC10-4F10-11
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Armenian Hamster
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR