Precision Sonic Sifter Sieves, 3", Advantec
Supplier: Advantec
L3-M5 L3-M100 L3-M70 L3-M55 L3-M80 L3-M60 L3-M20 L3-M40 L3-M65 L3-M85 L3-M90 L3-M125 L3-M105 L3-M25 L3-M45 L3-M50 L3-M75 L3-M150 L3-M15 L3-M95 L3-M10 L3-M30 L3-M35
57333-792EA 2672.08 CAD
57333-792 57333-794 57333-790 57333-800 57333-822 57333-812 57333-824 57333-802 57333-814 57333-796 57333-786 57333-830 57333-820 57333-798 57333-810 57333-788 57333-808 57333-826 57333-804 57333-816 57333-806 57333-828 57333-818
Precision Sonic Sifter Sieves, 3", Advantec
Sieves Test Sieves
These 3" diameter "see-through" acrylic framed sieves made with electroformed mesh are specifically designed to be used with the Sonic Sifter Separator for particles ranging from 3 µm to 150 µm
Made with electroformed precision nickel mesh meeting the current ASTM E161 sieve specifications.
Ordering information: Centerline® Sieve Inspection or Calibration Certification reports provide detailed statistical data that is linked to each serialized sieve and becomes an integral part of your Quality Assurance Program. Certifications are available as accessories.