Accessories for VWR® 12" Sieves
Supplier: VWR International
PB12HX CB12 PS12H PS12HX CS12 R8576 R8577 CB12W/R CS12W/R PB12F PB12FX PS12F PS12FX
47730-580EA 405.22 CAD
47730-580 47730-582 47730-584 47730-586 47730-588 47730-592 47730-594 10804-778 10804-780 10804-782 10804-784 10804-786 10804-788
Accessories for VWR® 12" Sieves
Sieves Test Sieves
VWR 12" diameter (304.8 mm) pans and covers fit all VWR 12" diameter sieves. Pans are half height (1-5/8") and are available in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available as accessories.