VWR® 12" Test Sieves, Intermediate Height, Brass and Stainless Steel
Supplier: VWR International
47730-038 47730-058 47730-066 47730-068 47730-070 47730-078 47730-082 47730-084 47730-086 47730-090 47730-092 47730-094 47730-096 47730-098 47730-102 47730-104 47730-106 47730-112 10811-528 10811-530 10811-532 10811-534 10811-590 10811-618 10811-620 10811-622 10811-624 10811-626 10811-628 10811-630 10811-632 10811-710 10811-712 10811-714 10811-716 10811-718 10811-720 10811-722 10811-724 10811-784 10811-808
47730-038EA 528.07 CAD
47730-038 47730-058 47730-066 47730-068 47730-070 47730-078 47730-082 47730-084 47730-086 47730-090 47730-092 47730-094 47730-096 47730-098 47730-102 47730-104 47730-106 47730-112 10811-528 10811-530 10811-532 10811-534 10811-590 10811-618 10811-620 10811-622 10811-624 10811-626 10811-628 10811-630 10811-632 10811-710 10811-712 10811-714 10811-716 10811-718 10811-720 10811-722 10811-724 10811-784 10811-808
VWR® 12" Test Sieves, Intermediate Height, Brass and Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Intermediate Height (2") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Brass and stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline®More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable