ProfiSampler®, Samplers, Bürkle
Supplier: BURKLE INC
5305-1000 5305-1100
10217-042EA 1781.69 CAD
10217-042 10217-050
ProfiSampler®, Samplers, Bürkle
Samplers Fluid Samplers
The ProfiSampler has been specially developed for rough industrial use.
  • Available with suction hose in PE, PA and PTFE wherever the highest degree of purity is required
  • Fill level control, automatic return flow and overfill protection through built-in venting valve
  • Sample bottles available in different materials and sizes
ProfiSampler Aluminium is used for solvents and flammable liquids and it consist of vacuum pump made of aluminium, with feet, venting valve and bottle thread GL 45 with glass bottle 500 ml, 5 suction tubes PA black, 220 cm long,More Product Information