Valve Brushes, Micronova
Supplier: Micronova
BRRE-40A BRWH-70 BRWH-40
10153-680EA 268.95 CAD
10153-680 76214-308 76415-984
Valve Brushes, Micronova
Brushes
The valve brush is a large, oval-shaped, polyester bristle brush with a plastic core and handle, designed for equipment cleaning.
  • Valve brush with attached handle
  • Ideal for cleaning drums, tanks, buckets
  • Polyester bristles, plastic handle
  • Available in a range of sizes
All materials of construction are produced from base resins. A model is available without handle for use with STA series handles.
Certifications: BPA-free and FDA approved.