Aluminum Extension Handles, Micronova
Supplier: Micronova
SSA-3 SSA-2
68400-168EA 130.32 CAD
68400-168 68400-742 89187-732 68400-307
Aluminum Extension Handles, Micronova
Mops
An ultra-lightweight, aluminum handle featuring three telescoping sections that extend to the desired length. Ideal for reaching high walls and ceilings, inside and around fixtures, and a range of cleanroom support applications.
  • Three telescoping sections for custom lengths
  • Ideal for extended reach in a range of cleanroom support applications
  • Available with extension from 6' to 18' (2 to 5.5 m)
  • Fits most mop frames and cleaning tools with threaded attachment sites
The Micronova Aluminum Extendible Mop Handles feature an ACME threaded end which fitsMore Product Information