Hand Held Scrapers, Micronova
Supplier: Micronova
HHSWH-10 HHSWH-8
89092-142CS 523.3 CAD
89092-142 89092-144
Hand Held Scrapers, Micronova
Scrapers
This hand held scraper tool is made with a seamless, one piece injection-mold that prevents bacterial growth. Designed for residue and build-up removal in clean and controlled environments.
  • Injection-molded, seamless construction
  • Will not scratch metal surfaces
  • Made from FDA approved polypropylene
  • Suitable for indirect food contact
  • Handle shaped for strong grip
  • Autoclavable
The scraper is chemical-resistant and will not rust or corrode.