Samplers for Viscous Media, ViscoSampler, Bürkle
Supplier: BURKLE INC
5331-1060 5331-1100 5331-1200 5331-2060 5331-2100 5331-2200
10217-148EA 1014.85 CAD
10217-148 10217-150 10217-344 10217-346 10217-348 10217-350
Samplers for Viscous Media, ViscoSampler, Bürkle
Samplers Fluid Samplers
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Specially developed for viscous substances such as slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions, creams etc.
  • No dirt accumulation
  • Disassembling and cleaning is very easy
  • Colour coded handles available
Made of transparent PP or ultra-pure and chemically inert PTFE/FEP for contamination-free sampling. The sampler can be disassembled and the non porous surfaces completely cleaned.