Sampler for Liquids, LiquiSampler, Bürkle
Supplier: BURKLE INC
5330-1060 5330-1100 5330-1200 5330-2060 5330-2100 5330-2200
10217-134EA 656 CAD
10217-134 10217-136 10217-138 10217-140 10217-142 10217-144
Sampler for Liquids, LiquiSampler, Bürkle
Samplers Fluid Samplers
Fluid sampler for easy, contamination-free sampling. Perfect for sampling from open and closed barrels, vats, tanks, silos, open waters (ponds, streams, lakes, rivers).
  • Disassembly and cleaning is simple
  • Easy to use, one hand push-button operation
  • Transparent
  • Suitable for point sampling, cross-sectional sampling, bottom sampling
Made of ultra-pure and chemically inert PTFE/FEP for contamination free sampling. Fits all commercially available vats and barrels with openings of at least 25 mm diameter. Easy to clean as all surfaces are free of pores andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable