QualiSampler Set, Bürkle
Supplier: BURKLE INC
5330-1000 5330-1001
10217-130EA 2282.52 CAD
10217-130 10217-132
QualiSampler Set, Bürkle
Samplers Fluid Samplers
The handy set, which comes complete with carrying case, combines the LiquiSampler and the ViscoSampler with practical accessories
Models in PP or PTFE/ FEP.
Ordering information: Set consists of LiquiSampler 100 cm, ViscoSampler 100 cm, case, 5 sample bottles and cleaning brush.