Liquid Sampler, Bürkle
Supplier: BURKLE INC
5326-0001 5326-0002 5326-0003 5326-0004
10217-122EA 820.6 CAD
10217-122 10217-124 10217-126 10217-128
Liquid Sampler, Bürkle
Samplers Fluid Samplers
Samplers for thin to viscous liquids.
  • Liquid Sampler, open with thumb: The valve is opened with the thumb to empty the sample
  • Liquid Sampler, place onto bottle: The valve is opened by attaching the sample container to empty the sample
Easy use with one-handed operation. With discharge funnel for safe filling of the sample bottles. Material V4A (1.4404) ; Container Diameter: 32 mm