LaboPlast Funnel Stands for Two or Four Funnels, Bürkle
Supplier: BURKLE INC
9622-0200 9622-0400
10217-298EA 324.9 CAD
10217-298 10217-300
LaboPlast Funnel Stands for Two or Four Funnels, Bürkle
Retort Stands
Sturdy and stable funnel stand, resistant to chemicals, shatter-proof and with continuously variable height adjustment.
  • For funnels Ø 40 - 180 mm (1.57 to 7.09")
  • Additional funnel holders are available as accessories
The funnel holders can be used with reducing inserts for small funnels. The funnel stand also makes an excellent filtering stand. Innovative magnet concept, no clamping screws are necessary. The height of the funnel holder can be adjusted easily and flexibly with one hand.
Packaging: One funnel stand per package.