Flow Indicators, Bürkle
Supplier: BURKLE INC
8720-1002 8720-1003 8720-1005 8720-2002 8720-2003 8720-2005
10217-286EA 35.07 CAD
10217-286 10217-288 10217-290 10217-292 10217-294 10217-296
Flow Indicators, Bürkle
Flow Indicators
These paddle wheel flow indicators give a clear visualisation of the flow rates of liquids or gas in tubing
LiquiMobil flow indicator made of SAN is mainly suitable for cooling systems and helps to reduce water consumption considerably because the cooling water fl ow can be adjusted better. Not suitable for aggressive substances. Maximum operating temperature LiquiMobil SAN is +30 °C.
CheMobil PMP shows reasonable chemical resistance, which makes it possible to use with a great number of fluids. CheMobil is not however appropriate for strong acids, bases and solvents. CheMobil is steam sterilisable. Maximum operating temperature +60 °C.
The flow indicator can operate in either direction of flow and can be mounted at any angle. The paddle wheel impeller starts to rotate at about 150 ml/min. Colour coding - red, black or blue