Foot-Operated Solvent Pumps, Bürkle
Supplier: BURKLE INC
5603-1100 5603-3000
10217-240EA 1830.7 CAD
10217-240 10217-244
Foot-Operated Solvent Pumps, Bürkle
Pumps Drum/Transfer Pumps
Solvent pump foot operated for filling of high purity liquids such as solvents. All medium-contacting parts are made of stainless steel (1.4301) and PTFE. Due to the barrel screw joint and the stopcock, the container and the pump can be hermetically sealed. Unwanted vapours can therefore not escape during the filling process. Massive, long-life industrial version. Clip connection for fast connection and disconnection of the air hose.
  • Immersion tube can be steplessly adjusted to the barrel depth
  • For barrels up to 220 litres
  • Suitable for steel barrels with R 2” thread
Accessories information: When decanting flammable liquids without the machines and containers involved in the process being sufficiently grounded, electrostatic build-up can occur, which for many substances can result in fire or an explosion. Burkle’s anti-static set (sold separately) ensures easy and reliable grounding. The anti-static set consists of three color-coded connecting copper cables with clamps in a handy plastic case.