Vials with Insert, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
225195 225200 225205 225190
89050-862CS 548.72 CAD
89050-862 89050-864 89050-866 66014-287
Vials with Insert, Wheaton
Vials
The glass/plastic vial with 0.1 ml insert has thermoplastic polymer outer shell provides safety from breakage.
The insert is made of Type I borosilicate glass. The conical interior of the insert ensures maximum retrival of contents without the hassle of using removable inserts.