Tubing Clamps, PVDF, Bürkle
Supplier: BURKLE INC
8619-0204 8619-0205 8619-0206 8619-0202 8619-0104 8619-0105 8619-0106 8619-0102 8619-0154 8619-0155 8619-0156 8619-0152
10217-272EA 19.2 CAD
10217-272 10217-274 10217-276 10217-382 10217-384 10217-386 10217-388 10217-390 10217-392 10217-394 10217-396 10217-398
Tubing Clamps, PVDF, Bürkle
Clamps Tubing and Sanitary Clamps
The micro adjustment screw enables the flow of liquids to be controlled delicately and smoothly or stopped completely.
  • PVDF with galvanised steel hook
  • Heat resistant up to +160 °C
  • Various sizes available for tubing from 1 - 20 mm external diameter
  • Available in four colours
The clamp can also be mounted on existing tube connections. The clamp's closure can be opened in any position, without screwing on and off.