LaboPlast® Pressure Sprayer, Bürkle
Supplier: BURKLE INC
0309-0100 0309-0120 0309-0130
10216-878EA 133.41 CAD
10216-878 76501-520 76501-522
LaboPlast® Pressure Sprayer, Bürkle
Bottles Spray Bottles
Pressure sprayer used for disinfection, cleaning, plant protection, thin layer chromatography, technical and sanitary use. The hand pump is used to produce a pressure of up to 4 bar in the container. Spraying take place by pressing a button.
  • Media-contacting parts PP, PE, glass-fibre-reinforced PA, V2A, FKM
  • Bottle made of HDPE with nozzle diameter 0.6 mm (0.02")
  • Adjustable spray jet
  • Temperature max. 30 °C