Culture Vials with Cap, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
225145 225142 225144
66014-242CS 303.47 CAD
66014-242 66014-235 66014-240
Culture Vials with Cap, WHEATON®, DWK Life Sciences
Vials
The autovclavable culture vials feature deep-skirted screw caps to allow safer, more dependable handling when working with infectious materials.
These vials are manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements. The pre-attached deep skirted solid-top black phenolic screw cap with 14B rubber liner.
Order Now
SPECIFICATIONS
Environmentally Preferable