VWR® Plate Centrifuge
Supplier: VWR
VWR®
C2001-VWR
10144-214EA 2044.83 CAD
10144-214
VWR® Plate Centrifuge
Centrifuges Microcentrifuges
Spins at 2500 rpm/ 850xg, to quickly spin down droplets and condensation in PCR plates.
  • Easy access rotor includes two plate carriers
  • Unique swing-out rotor design prevents sample spillage
  • Less than half the size of traditional centrifuges
Accepts all popular PCR plates: skirted, half-skirted and non-skirted. Gray color.
Certifications: CE marked.
Ordering information: Supplied with adapter for non-skirted plates.