Coliwasa Trumpet, Disposable, Wheaton®
Supplier: DWK Life Sciences
Coliwasa Trumpet
885235
66174-235CS 660.31 CAD
66174-235
Coliwasa Trumpet, Disposable, Wheaton®
Samplers Disposable Samplers
Use for sampling sludge or semi-solid material.
Disposable glass coliwasa will bore through semi-solid sludge type samples. Liquid and sludge material will remain in the coliwasa due to its tight PTFE resin seal. Tube is 101.6 cm long with 200 ml capacity.