Anti-IFNG Rat Monoclonal Antibody (Biotin) [clone: XMG1.2]

Supplier: Tonbo Biosciences
30-7311-U025 30-7311-U100
10208-348EA 64.78 CAD
10208-348 10208-350
Anti-IFNG Rat Monoclonal Antibody (Biotin) [clone: XMG1.2]
Antibodies
The XMG1.2 antibody is specific for mouse Interferon-gamma (IFN-g), a 20 kDa type II cytokine known for its central roles in protection against bacterial or viral pathogens and for its anti-tumor properties. IFN-g is secreted by several types of immune cells which allow the cytokine to modulate innate immunity when secreted by NK and NKT cells, and to function in support of adaptive immunity when secreted by Th1 and CD8+ T cells (CTLs).

The XMG1.2 antibody is suitable for detection of intracellular IFN-g protein by flow cytometry. This format can be used for quantitative analysis of the secreted protein by ELISA when paired with an appropriate capture antibody. This clone has been reported for neutralization of the functional activity of IFN-g in a variety of assays. Please choose the appropriate format for each application.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: XMG1.2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR