Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT2]

Supplier: Tonbo Biosciences
20-0389-T025 20-0389-T100
10208-420EA 74.26 CAD
10208-420 10208-422
Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT2]
Antibodies
The HIT2 antibody is specific for human CD38, a 45 kD type II transmembrane glycoprotein expressed on thymocytes, activated T and B cells, and monocytes as well as other non-hematopoietic cells. CD38 is an ectoenzyme that functions to catalyze the synthesis and hydrolysis of cyclic ADP-ribose and is involved in cell signaling, regulating cell adhesion, and activation. Additionally, CD38 has been shown to be a prognostic marker for some leukemias and other diseases.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR