LaboPlast® Spray Bottles, Turn 'n' Spray with Overhead Valve, Bürkle
Supplier: BURKLE INC
0309-0002 0309-0005 0309-0010
10216-872EA 25.93 CAD
10216-872 10216-874 10216-876
LaboPlast® Spray Bottles, Turn 'n' Spray with Overhead Valve, Bürkle
Bottles Spray Bottles
Spraying upside down is not possible with standard spray bottles. As soon as the suction tube is no longer immersed in liquid, it cannot spray. Not so with the overhead spray bottle Turn 'n' Spray. It features a special technology that allows you to spray from any position. That means it can spray practically and conveniently from any direction and reach hard to access places easily and without straining.
  • Uninterrupted spraying, even when inverted
  • Adjustable nozzle, diameter 0.6 mm
  • Amount sprayed per stroke: 1.2 mL ± 0.1
  • LDPE, translucent, PP spray
More Product Information