Percoll®, yellow liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219536925 0219536990
IC0219536925EA 120.82 CAD
IC0219536925 IC0219536990
Percoll®, yellow liquid
Percoll®

Percoll® is a medium for density gradient centrifugation of cells, viruses and subcellular particles. Percoll® is composed of colloidal silica coated with polyvinylpyrrolidone (PVP), which renders the material completely non-toxic and ideal for use with biological materials.Centrifugation of Percoll® results in spontaneous formation of a density gradient due to the heterogeneity of particle sizes in the medium. Percoll® can be used for formation of gradients either by the use of conventional gradient mixers or by high speed centrifugation. In the latter case the sample can be premixed with the medium and then separated on the gradient created in situ. In this way gradient formation and sample separation can be achieved in a single operation.

Storage Temperature: Refrigerator
MDL Number: MFCD00131830
CAS Number: 65455-52-9

Order Now

Specification Test Results

Identity Test Passes
Viscosity Record cp at 20 deg C
Osmolality Max 25 mOsm/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR