D-(+)-Sucrose ≥95%, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranosideSucroseD(+)-Saccharoseβ-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranosideα-ᴅ-Glc-(1→2)-β-ᴅ-Fru

0219027101 02190271.5 0219027105
IC190271.5EA 121.47 CAD
IC19027101 IC19027105 IC190271.5
D-(+)-Sucrose ≥95%, crystalline powder
D(+)-Sucrose

Soluble in water (1 g/0.5 mL at 25 °C; 1 g/0.2 mL boiling water), ethanol (1 g/170 mL) or methanol (1 g/100 mL); moderately soluble in glycerol or pyridine.


Sucrose can be used to prepare density gradients for cell/organelle separation. The refractive index of the solution will be directly related to the density. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays.

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
MDL Number: MFCD00006626
CAS Number: 57-50-1
https://ca.vwr.com/stibo/web/std.lang.all/12/10/36421210.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR