Starch, soluble, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: StarchVitex® iodine indicatorAmylodextrin

0215258290 0215258280 0215258291
IC15258280EA 95.73 CAD
IC15258280 IC15258291 IC15258290
Starch, soluble, powder ACS
Starch, soluble

Soluble in water.


Starch is used as an additive for food processing, food starches are typically used as thickeners and stabilizers. Starch is used to produce various bioplastics, synthetic polymers that are biodegradable. An example is polylactic acid. Glucose from starch can be further fermented to biofuel corn ethanol. Hydrogen production can use starch as the raw material.

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
MDL Number: MFCD00082026
CAS Number: 9005-84-9

Order Now

Specification Test Results

Form White Powder
Storage Store at Room Temperature(15-30 °C).
pH 5.0 - 7.0(2% aq soln)
Residue on Ignition ≤0.4%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR