Lecithin (from soy bean), dark yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210214780 0210214783 0210214790 0210214791
IC10214780EA 106.32 CAD
IC10214780 IC10214783 IC10214790 IC10214791
Lecithin (from soy bean), dark yellow powder
Lecithin

Lecithin from Soy, Refined, is a natural mixture of phospholipids consisting of phosphatidyl choline, phosphatidyl ethanolamine, and phosphatidyl inositol. In soybean lecithin the predominant fatty acid is polyunsaturated linoleic acid, amounting to about 70% of the fatty acids present.
Lecithin is used for preparation of vesicle suspensions commonly called liposomes or as monolayers.
A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.


  • Presentation: Dark Yellow Powder
  • Storage temperature: Store at -0 °C.


Acetone Insolubles 95%


Lecithin from Soy, Refined, is a natural mixture of phospholipids consisting of phosphatidyl choline, phosphatidyl ethanolamine, and phosphatidyl inositol. In soybean lecithin the predominant fatty acid is polyunsaturated linoleic acid, amounting to about 70% of the fatty acids present.

Storage Temperature: Freezer
MDL Number: MFCD00147406
CAS Number: 8002-43-5

Order Now

Specification Test Results

Identity Test Passes
Source Soybean (refined oil removed)
Moisture ≤2%
Acetone Insolubles ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR