Microtest Manifolds, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
851381 851388
53500-336EA 1291.7 CAD
53500-336 53500-343
Microtest Manifolds, DWK Life Sciences
Manifolds
These autoclavable stainless steel microtest manifolds are ideal for use with 24 or 96 well microplates
It tightly fit with both Luer and Luer Lock syringe nozzles. The manifolda available in both a straight "T" style and an angular style with an offset hub.
Order Now
SPECIFICATIONS
Environmentally Preferable