Seals for Headspace Vials, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W224215 W224216 W224217 W224221 W224224 W224225 W224223
14227-784CS 389.76 CAD
14227-784 14227-786 14227-788 89495-256 89495-258 89495-260 89495-262
Seals for Headspace Vials, Wheaton, DWK Life Sciences
Closures Chromatography Vial Closures
Pressure release seals feature score lines that allow for internal pressure release when 3.0 ±0.5 bar has been exceeded.
Order Now
SPECIFICATIONS
Environmentally Preferable