SP Bel-Art ProCulture Micro-Tube Homogenizer System, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F65000-0002 F65000-0005 F65000-0006 F65000-9001 F65000-9002 F65000-9006 F65100-0000
89524-800EA 184.89 CAD
89524-800 89524-802 89524-804 89524-806 89524-808 89524-810 89524-812 89524-814 89524-816 89524-820 89524-822 89524-824 76529-604
SP Bel-Art ProCulture Micro-Tube Homogenizer System, Bel-Art Products, a part of SP
Homogenizers Handheld Homogenizers
Hand-held, cordless homogenizer features a simple push in / pull out pestle exchange and is ideal for thorough and faster processing of one to many samples.
  • Pestles and tubes assure tight tolerances and thorough homogenization and grinding of materials resulting in optimal efficiency of pellet reconstitution or re-suspension
  • Pestles and tubes are chemical and heat resistant; colorant-free to eliminate potential leaching of heavy metals into samples
  • Tubes have 25% thicker bottoms than industry standard to ensure integrity during use
  • Each tube has frosted
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable