Chromatography Gravity Columns with Standard Taper Joint and Fritted Disk, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
420620-0120 420620-0130 420620-0320 420620-0425 420620-0660 420620-0750 420620-0760
KT420620-0120EA 468.45 CAD
KT420620-0120 KT420620-0130 KT420620-0320 KT420620-0425 KT420620-0660 KT420620-0750 KT420620-0760
Chromatography Gravity Columns with Standard Taper Joint and Fritted Disk, Kimble Chase
Chromatography Columns
Gravity chromatography column manufactured from medium wall tubing and in-line reservoir for containing liquid buffer.
Standard Taper joint size 24/40. A coarse porosity glass frit (40 to 60 µm pore size) is a support for column packing material. Custom sizes and plastic safety coating are available upon request.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: PTFE stopcock needs to be purchased separately.