Autoclavable Precision Clean® Breather Bags, PPC Flexible Packaging
Supplier: Fisher Container
STCK0041-001 STCK0042-001 STCK0043-001 STCK0044-001 STCK0045-001 STCK0047-001 STCK0046-001
89496-126CS 2834.03 CAD
89496-126 89496-128 89496-130 89496-132 89496-134 89496-136 89496-138
Autoclavable Precision Clean® Breather Bags, PPC Flexible Packaging
Bags Sterilization Pouches
A combination of Tyvek™ and clear, ultraclean film structure aids in the prevention of interior particle contamination.
  • Faster cycle times
  • Molecular bonded seals
  • Pouch suitable for sterilizaiton by steam, EtO, gamma radiation or hydrogen peroxide
  • One side Tyvek™ and reverse side Precision Clean® HD 3-mil clear film
It is designed for products to be used in cleanroom environments that also require sterilization.
Certifications: Certified to NASA™ clean standards by Fisher to meet Level 100 KSC-C-123.
Order Now
SPECIFICATIONS
Environmentally Preferable