Chromatography Glass Columns with Reservoir, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
420610-0125 420610-0320 420610-0430 420610-0450 420610-0530 420610-0550 420610-0560 420610-0630 420610-0660 420610-0730
KT420610-0125EA 434.07 CAD
KT420610-0125 KT420610-0320 KT420610-0430 KT420610-0450 KT420610-0530 KT420610-0550 KT420610-0560 KT420610-0630 KT420610-0660 KT420610-0730
Chromatography Glass Columns with Reservoir, Kimble Chase
Chromatography Columns
Gravity chromatography column manufactured from medium wall tubing with PTFE stopcock and in-line reservoir for containing liquid buffer.
  • Standard taper joint size 24/40
Glass wool (not supplied) may be used as a support for the packing material. Custom sizes and plastic safety coating are available upon request.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.