Celstir® Suspension Culture Flasks, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
356987 356984
25461-038EA 3602.28 CAD
25461-038 25461-040
Celstir® Suspension Culture Flasks, Wheaton
Flasks Spinner Flasks
Four arm flask with fluoropolymer resin lined sidearm caps.
  • Even stirring action
  • Each sidearm measures 45 mm
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable