Snap Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
242775 242786 242776 242772
66014-612CS 2137.74 CAD
66014-612 89235-548 89235-556 89235-566
Snap Caps, DWK Life Sciences
Closures Chromatography Vial Closures
Eliminate the need for crimping and decapping tools.
  • Manufactured from polyethylene
Fits the WHEATON E-Z vials® with snap ring top.
Order Now
SPECIFICATIONS
Environmentally Preferable