Flow Check Airflow Indicator, Draeger
Supplier: Draeger
6400761 6400812
CA13450-860EA 5372.54 CAD
CA13450-860 CA13450-864
Flow Check Airflow Indicator, Draeger
Gas Detection Systems
A small heating element in the device head heats an alcohol mixture with a high molecular weight and density similar to air
If the liquid comes into contact with air, it condenses into a type of mist. The source, flow and speed of the slightest air streams can be identified with the naked eye in this way.
Ordering information: Supplied in a black plastic carrying case with a rechargeable battery pack and a pack of three mist cartridges. There is also plenty of space in the carrying case for the battery charger, spare battery and the adapter for the in-car cigarette lighter. The charging device must be ordered separately.