Safety Coated Reagent Bottle, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
221014 221017
16150-080CS 225.3 CAD
16150-080 16150-082
Safety Coated Reagent Bottle, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Safety coated narrow neck reagent bottle for the safe storage of laboratory samples.
  • Plastisol coated exterior of bottle retains contents if bottle is accidentally broken
  • Made from low extractable clear borosilicate glass
  • Non-slip coating allows for greater ease and stability when handling wet or dry
  • Supplied with attached white polypropylene 33-430 cap with PTFE liner
  • Autoclavable
Certifications: It conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable